missing translation for 'onlineSavingsMsg'
Learn More

p-Salicylic Acid, TRC

Product Code. 17952545
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17952545 100 g 100g
17913895 10 g 10g
17931965 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17952545 Supplier Toronto Research Chemicals Supplier No. TRCS088120100G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-96-7
Molecular Weight (g/mol) 138.12
Synonym 4-Hydroxybenzoic Acid, Ethyl Parahydroxybenzoate Imp. A (EP), Butyl Parahydroxybenzoate Imp. A (EP), Propyl Parahydroxybenzoate Imp. A (EP), Methyl Parahydroxybenzoate Imp. A (EP), Acetylsalicylic Acid Imp. A (EP), Salicylic Acid Imp. A (EP), 4-Carboxyphenol, 4-Hydroxybenzoic acid, NSC 4961, Paraben-acid, p-Carboxyphenol, p-Hydroxybenzoic acid, p-Salicylic acid
IUPAC Name 4-hydroxybenzoic acid
SMILES OC(=O)c1ccc(O)cc1

Specifications

Molecular Formula C7 H6 O3
Quantity 100 g
InChI Formula InChI=1S/C7H6O3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H,(H,9,10)
Formula Weight 138.0317
Percent Purity >95%
Chemical Name or Material 4-Hydroxybenzoic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.