missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p-Tolualdehyde-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 4-Methylbenzaldehyde-2,3,5,6-D4 |
| CAS | 1219804-07-5 |
| Molecular Formula | C8 D4 H4 O |
| Quantity | 0.25 g |
| Synonym | p-Tolualdehyde-2,3,5,6-d4, p-Tolualdehyde-D4, 4-Methylbenzaldehyde-d4, 4-Methylbenzaldehyde-2,3,5,6-d4, 4-Tolualdehyde-d4, 4-Methylbenzaldehyde-2,3,5,6-D4 |
| InChI Formula | InChI=1 S/C8H8O/c1-7-2-4-8(6-9)5-3-7/h2-6 H,1H3/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(C=O)c([2 H])c([2 H])c1C |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-methylbenzaldehyde |
| Molecular Weight (g/mol) | 124.173 |
| Formula Weight | 124.083 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction