missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfinic acid, sodium salt, 97%, anhydrous

Product Code. 11945751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11945751 100 g Glass bottle 100g
11935751 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11945751 Supplier Thermo Scientific Acros Supplier No. 434531000
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 824-79-3 | C7H7O2S·Na | 178.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-79-3
Molecular Formula C7H7O2S·Na
Molecular Weight (g/mol) 178.18
InChI Key KFZUDNZQQCWGKF-UHFFFAOYSA-M
Synonym sodium 4-methylbenzenesulfinate, sodium p-toluenesulfinate, p-toluenesulfinic acid sodium salt, sodium toluene-4-sulphinate, sodium p-tolylsulfinate, sodium 4-toluenesulfinate, benzenesulfinic acid, 4-methyl-, sodium salt, unii-7cku5w4trm, p-toluenesulfinic acid, sodium salt, p-toluensulfinan sodny czech
PubChem CID 2723791
IUPAC Name sodium;4-methylbenzenesulfinate
SMILES CC1=CC=C(C=C1)S(=O)[O-].[Na+]

Specifications

Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: soluble.
Formula Weight 178.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material p-Toluenesulfinic acid, sodium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.