missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonic acid monohydrate, 99%, extra pure

Product Code. 10356830 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Unit Size:
100g
10kg
2.5kg
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10356830 500 g Plastic bottle 500g
10396780 5 g Glass Bottle 5g
10455993 100 g Plastic bottle 100g
10380004 2.5 kg Plastic bottle 2.5kg
11924561 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10356830 Supplier Thermo Scientific Acros Supplier No. 139025000
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
This item is not returnable. View return policy

CAS: 6192-52-5 | C7H10O4S | 190.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6192-52-5
Molecular Formula C7H10O4S
Molecular Weight (g/mol) 190.213
MDL Number MFCD00142137
InChI Key KJIFKLIQANRMOU-UHFFFAOYSA-N
Synonym p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
PubChem CID 521998
IUPAC Name 4-methylbenzenesulfonic acid;hydrate
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.O

Specifications

Melting Point 100.0°C to 108.0°C
Assay Percent Range 99%
Linear Formula CH3C6H4SO3H·H2O
Fieser 01,1172; 04,508; 05,673; 06,597; 07,374; 08,488; 09,471; 11,535; 12,507
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether and chloroform
Formula Weight 190.22
Percent Purity 99%
Grade Extra Pure
Quantity 500 g
Flash Point 180°C
Packaging Plastic bottle
Chemical Name or Material p-Toluenesulfonic acid monohydrate, >98.5% (total TSA)
Show More Show Less
compliance-icons
Product Identifier
  • p-Toluenesulfonic acid monohydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.