missing translation for 'onlineSavingsMsg'
Learn More

p-Toluenesulfonic acid sodium salt, 90+%

Product Code. 11449586
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449586 50 g 50g
11453937 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449586 Supplier Thermo Scientific Alfa Aesar Supplier No. A19332.18
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 657-84-1 | C7H7NaO3S | 194.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 657-84-1
Molecular Formula C7H7NaO3S
Molecular Weight (g/mol) 194.18
MDL Number MFCD00798566,MFCD00064388
InChI Key KVCGISUBCHHTDD-UHFFFAOYSA-M
Synonym sodium p-toluenesulfonate, sodium 4-methylbenzenesulfonate, sodium tosylate, p-toluenesulfonic acid sodium salt, sodium toluenesulfonate, sodium toluenesulphonate, tosylate, sodium, naxonate hydrotrope, cyclophil sts 70, eltesol st 34
PubChem CID 3720192
IUPAC Name sodium;4-methylbenzenesulfonate
SMILES [Na+].CC1=CC=C(C=C1)S([O-])(=O)=O

Specifications

Merck Index 14,9533
Formula Weight 194.19 (Anhydrous)
Percent Purity ≥90%
Quantity 50 g
Chemical Name or Material p-Toluenesulfonic acid sodium salt
compliance-icons
Product Identifier
  • p-Toluenesulfonic acid sodium salt hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: p-Toluenesulfonic acid sodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.