missing translation for 'onlineSavingsMsg'
Learn More

p-Tolunitrile, 98+%

Product Code. 10554091 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
250g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10554091 250 g Glass bottle 250g
10510522 5 g Glass bottle 5g
10544091 100 g Glass bottle 100g
10317430 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10554091 Supplier Thermo Scientific Acros Supplier No. 158022500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-85-8 | C8H7N | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-85-8
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.15
MDL Number MFCD00001827
InChI Key VCZNNAKNUVJVGX-UHFFFAOYSA-N
Synonym p-tolunitrile, 4-cyanotoluene, benzonitrile, 4-methyl, 4-tolunitrile, p-methylbenzonitrile, p-toluonitrile, p-cyanotoluene, p-tolylnitrile, p-tolunitril, p-toluenenitrile
PubChem CID 7724
IUPAC Name 4-methylbenzonitrile
SMILES CC1=CC=C(C=C1)C#N

Specifications

Melting Point 26.0°C to 28.0°C
Density 0.9800g/mL
Boiling Point 103.0°C to 106.0°C (20.0 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CN
Refractive Index 1.5285 to 1.5305
Quantity 250 g
Beilstein 09, 489
Merck Index 15, 9698
Specific Gravity 0.98
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 117.15
Percent Purity 98+%
Physical Form Viscous Liquid
Chemical Name or Material p-Tolunitrile, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.