missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyl isothiocyanate, 97%

Product Code. 11450413
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450413 100 g 100g
11430413 5 g 5g
11440413 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450413 Supplier Thermo Scientific Alfa Aesar Supplier No. A11499.22
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 622-59-3 | C8H7NS | 149.211 g/mol

p-Tolyl isothiocyanate has been used in the preparation of 6-[1-amino-3-(p-tolyl)-thiourea]-2-ethylbenzo[de]isoquinoline-1,3-dione. It is used as capping agent to cap the N-terminal ends of polypeptide. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolyl isothiocyanate has been used in the preparation of 6-[1-amino-3-(p-tolyl)-thiourea]-2-ethylbenzo[de]isoquinoline-1,3-dione. It is used as capping agent to cap the N-terminal ends of polypeptide. It is also used as pharmaceutical intermediate.

Solubility
Decompose in water.

Notes
Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-59-3
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.211
MDL Number MFCD00004813
InChI Key ABQKHKWXTUVKGF-UHFFFAOYSA-N
Synonym p-tolyl isothiocyanate, 4-methylphenyl isothiocyanate, 4-methylphenylisothiocyanate, p-tolylisothiocyanate, 4-tolylisothiocyanate, benzene, 1-isothiocyanato-4-methyl, 4-tolyl isothiocyanate, isothiocyanic acid, p-tolyl ester, 1-isothiocyanato-4-methyl-benzene, isothiocyanic acid, 4-tolyl ester
PubChem CID 12149
IUPAC Name 1-isothiocyanato-4-methylbenzene
SMILES CC1=CC=C(C=C1)N=C=S

Specifications

Melting Point 23°C to 26°C
Boiling Point 236°C to 238°C
Flash Point 107°C (224°F)
Odor Pungent
Refractive Index 1.6345
Quantity 100 g
UN Number UN2923
Beilstein 386032
Sensitivity Moisture sensitive
Solubility Information Decompose in water.
Formula Weight 149.22
Percent Purity 97%
Chemical Name or Material p-Tolyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.