missing translation for 'onlineSavingsMsg'
Learn More

p-Tolylacetylene, 98%

Product Code. 11496566
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11496566 5 g 5g
11406576 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11496566 Supplier Thermo Scientific Alfa Aesar Supplier No. B24812.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 766-97-2 | C9H8 | 116.163 g/mol

4-Ethynyltoluene was used in the synthesis of bis(bicyclic) products by the cycloaddition of alkynes to the AlN2C2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethynyltoluene was used in the synthesis of bis(bicyclic) products by the cycloaddition of alkynes to the AlN2C2.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, heavy metal powders.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-97-2
Molecular Formula C9H8
Molecular Weight (g/mol) 116.163
MDL Number MFCD00008571
InChI Key KSZVOXHGCKKOLL-UHFFFAOYSA-N
Synonym 4-ethynyltoluene, p-tolylacetylene, 4-methylphenylacetylene, p-ethynyltoluene, p-methylphenylacetylene, benzene, 1-ethynyl-4-methyl, toluene, p-ethynyl, benzene, 1-ethynyl-4-methyl-9ci, 4'-methylphenyl acetylene, 1-ethynyl-4-methyl-benzene
PubChem CID 13018
IUPAC Name 1-ethynyl-4-methylbenzene
SMILES CC1=CC=C(C=C1)C#C

Specifications

Melting Point ∼21°C
Density 0.916
Boiling Point 168°C to 170°C
Flash Point 49°C (120°F)
Odor Pungent
Refractive Index 1.547
Quantity 5 g
UN Number UN3295
Beilstein 969653
Solubility Information Insoluble in water.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material p-Tolylacetylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.