missing translation for 'onlineSavingsMsg'
Learn More

p-Tolyltrichlorosilane, 97%, Thermo Scientific Chemicals

Product Code. 11430337
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430337 5 g 5g
11440337 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430337 Supplier Thermo Scientific Alfa Aesar Supplier No. B21728.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

It can be used to produce 3-(p-Methylphenyl)cyclohexanon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used to produce 3-(p-Methylphenyl)cyclohexanon.

Solubility
Reacts with water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 701-35-9
Molecular Formula C7H7Cl3Si
Molecular Weight (g/mol) 225.568
MDL Number MFCD00045131
InChI Key WOMUGKOOLXQCTQ-UHFFFAOYSA-N
Synonym p-tolyltrichlorosilane, tolyltrichlorosilane, trichloro-p-tolylsilane, silane, trichloro 4-methylphenyl, trichloro 4-methylphenyl silane, trichloro p-tolyl silane, silane, trichloro-p-tolyl, benzene, 1-methyl-4-trichlorosilyl, 4-tolyltrichlorosilane, p-tolylsilyl trichloride
PubChem CID 69697
IUPAC Name trichloro-(4-methylphenyl)silane
SMILES CC1=CC=C(C=C1)[Si](Cl)(Cl)Cl

Specifications

Boiling Point 218°C to 220°C
Quantity 5 g
UN Number UN2987
Beilstein 1866557
Flash Point 98°C (208°F)
Solubility Information Reacts with water.
Formula Weight 225.58
Percent Purity 97%
Odor Irritating
Refractive Index 1.524
Sensitivity Moisture sensitive
Density 1.28
Chemical Name or Material p-Tolyltrichlorosilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.