missing translation for 'onlineSavingsMsg'
Learn More

p-Vinylguaiacol, TRC

Product Code. 30004808
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004808 25 g 25g
17921665 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004808 Supplier Toronto Research Chemicals Supplier No. TRCV42496025G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7786-61-0
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.17
Synonym 4-Ethenyl-2-methoxy-phenol, 4-Vinyl-guaiacol, 2-Methoxy-4-vinyl-phenol, (4-Hydroxy-3-methoxyphenyl)ethene, 2-Methoxy-4-ethenylphenol, 2-Methoxy-4-vinylphenol, 3-Methoxy-4-hydroxystyrene, 4-Ethenyl-2-methoxyphenol, 4-Hydroxy-3-methoxystyrene, 4-Hydroxy-3-methoxyvinylbenzene, 4-Vinyl-2-methoxyphenol, 4-Vinylguaiacol, Varamol 106, o-Methoxy-p-vinylphenol;
IUPAC Name 4-ethenyl-2-methoxyphenol
SMILES OC1=CC=C(C=C)C=C1OC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H10O2/c1-3-7-4-5-8(10)9(6-7)11-2/h3-6,10H,1H2,2H3
Formula Weight 150.0681
Percent Purity >95%
Chemical Name or Material p-Vinylguaiacol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.