missing translation for 'onlineSavingsMsg'
Learn More

p-Xylene, ≥99% (GC), Solstice

Product Code. 15677560
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15677560 2.5 L Glass Bottle 2.5 L
15667560 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15677560 Supplier Fluka Supplier No. 956822.5L
  • ONLINE EXCLUSIVE PRICE
    252.00 €
    237.04 € / 2.5 L
    Save 14.96 €
    5% Off
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 106-42-3 | C8H10 | 106.17 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-42-3
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008556
InChI Key URLKBWYHVLBVBO-UHFFFAOYSA-N
Synonym p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
PubChem CID 7809
ChEBI CHEBI:27417
IUPAC Name 1,4-xylene
SMILES CC1=CC=C(C)C=C1

Specifications

Melting Point 12-13°C
Density 0.861g/mL at 20°C
Quantity 2.5 L
Explosion Limit 0.07
Flash Point 25°C (77°F)
Ignition Point 984°F
Packaging Glass Bottle
Refractive Index 1.497
Linear Formula C6H4(CH3)2
Vapor Pressure 9mmHg (20°C)
UN Number UN1307
Beilstein 1901563
Boiling Point 138°C to 139°C
Formula Weight 106.17g/mol
Percent Purity ≥99.0% (GC)
Grade Not Available
Chemical Name or Material p-Xylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.