missing translation for 'onlineSavingsMsg'
Learn More

p-Xylene, CHROMASOLV™, for HPLC, ≥99%, Honeywell Riedel-de Haën™

Product Code. 15663510
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15663510 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15663510 Supplier Honeywell-Chromasolv Supplier No. 3171951L
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CHROMASOLV™, for HPLC,≥99%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 12°C to 13°C (literature)
Absorbance max. 0.01 at 400 nm, max. 0.01 at 380 nm, max. 0.05 at 350 nm, max. 0.10 at 320 nm, max. 1.0 at 294 nm
CAS 106-42-3
CAS Max % 100.0000%
Density 0.861 g/mL at 20 °C(lit.)
Quantity 1 L
Flash Point 25°C
Packaging Glass Bottle
Molecular Formula C8H10
Linear Formula C6H4(CH3)2
MDL Number MFCD00008556
UN Number UN1307
Beilstein 1901563
Synonym p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
Boiling Point 137°C to 139°C
InChI Key URLKBWYHVLBVBO-UHFFFAOYSA-N
SMILES CC1=CC=C(C)C=C1
IUPAC Name 1,4-xylene
Molecular Weight (g/mol) 106.17
PubChem CID 7809
ChEBI CHEBI:27417
Formula Weight 106.17
Percent Purity >99%
Grade HPLC
Chemical Name or Material p-Xylene, CHROMASOLV™, for HPLC, ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.