missing translation for 'onlineSavingsMsg'
Learn More

P-Xylene, Pure, Fisher Chemical

Product Code. 10477460 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10477460 500 mL Amber Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10477460 Supplier Fisher Chemical Supplier No. X/0400/08
Quantity Price
16 78.95 € / 500mL
1 85.35 € / 500mL
4 83.10 € / 500mL

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Specifications

Melting Point 13°C
CAS 106-42-3
Color Colorless
Quantity 500 mL
Packaging Amber Glass Bottle
Physical Form Liquid
Molecular Formula C8H10
Vapor Pressure 8mbar at 20°C
MDL Number MFCD00008556
Viscosity 0.648 mPaS ( at 20°C)
Synonym p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
Boiling Point 138°C
InChI Key URLKBWYHVLBVBO-UHFFFAOYSA-N
SMILES CC1=CC=C(C)C=C1
IUPAC Name 1,4-xylene
Molecular Weight (g/mol) 106.17
PubChem CID 7809
ChEBI CHEBI:27417
Formula Weight 106.17g/mol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • p-Xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.