Product Code. 15631330
Shop All Solstice Research Chemicals Products
Click to view available options
Quantity:
500 mL
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
2.5 L
20 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
15631330
500 mL
Glass Bottle
500mL
15611330
2.5 L
Glass Bottle
2.5 L
15621330
20 L
Stainless Steel Can
20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
15631330
Supplier
Honeywell Research Chemicals
Supplier No.
134449500ML
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Reagent Grade, 99%
≥95% for general laboratory use.
Chemical Identifiers
CAS
106-42-3
Molecular Formula
C8H10
Molecular Weight (g/mol)
106.17
MDL Number
MFCD00008556
InChI Key
URLKBWYHVLBVBO-UHFFFAOYSA-N
Synonym
p-xylene, para-xylene, 1,4-dimethylbenzene, p-methyltoluene, p-dimethylbenzene, p-xylol, benzene, 1,4-dimethyl, 4-xylene, chromar, 4-methyltoluene
PubChem CID
7809
ChEBI
CHEBI:27417
IUPAC Name
1,4-xylene
SMILES
CC1=CC=C(C)C=C1
Specifications
Melting Point
12°C
Color
Colorless
Density
0.861g/mL at 20°C
Quantity
500 mL
Explosion Limit
0.07
Flash Point
25°C (77°F)
Ignition Point
984°F
Packaging
Glass Bottle
Physical Form
Liquid
Refractive Index
n20/D 1.495(lit.)
Linear Formula
C6 H4 (CH3 )2
Vapor Pressure
9mmHg (20°C)
UN Number
UN1307
Beilstein
1901563
Boiling Point
137°C to 139°C
Solubility Information
Soluble in water, 0.2g/L
Formula Weight
106.17g/mol
Percent Purity
99%
Grade
ReagentPlus
Chemical Name or Material
P-Xylene
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?