Product Code. 30454022
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Unit Size
30454022
1 L
1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
30454022
Supplier
Toronto Research Chemicals
Supplier No.
TRCX7494101L
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Specifications
Chemical Name or Material
p-Xylene (1,4-Dimethylbenzene)
CAS
106-42-3
Molecular Formula
C8 H10
Quantity
1 L
Synonym
Benzene','1,4-dimethyl-, p-Xylene (8CI), 1,4-Dimethylbenzene, 1,4-Xylene, 4-Methyltoluene, NSC 72419, p-Dimethylbenzene, p-Methyltoluene, p-Phenylenebis(methylene), p-Xylol
InChI Formula
InChI=1S/C8H10/c1-7-3-5-8(2)6-4-7/h3-6H,1-2H3
SMILES
Cc1ccc(C)cc1
IUPAC Name
1,4-xylene
Molecular Weight (g/mol)
106.16
Formula Weight
106.0783
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?