missing translation for 'onlineSavingsMsg'
Learn More

p-Xylene, TRC

Product Code. 30455148
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455148 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 30455148 Supplier Toronto Research Chemicals Supplier No. TRCX749410500ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material p-Xylene (1,4-Dimethylbenzene)
CAS 106-42-3
Molecular Formula C8 H10
Quantity 500 mL
Synonym Benzene','1,4-dimethyl-, p-Xylene (8CI), 1,4-Dimethylbenzene, 1,4-Xylene, 4-Methyltoluene, NSC 72419, p-Dimethylbenzene, p-Methyltoluene, p-Phenylenebis(methylene), p-Xylol
InChI Formula InChI=1S/C8H10/c1-7-3-5-8(2)6-4-7/h3-6H,1-2H3
SMILES Cc1ccc(C)cc1
IUPAC Name 1,4-xylene
Molecular Weight (g/mol) 106.16
Formula Weight 106.0783
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.