missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Microplate BCA Protein Assay Kit - Reducing Agent Compatible

Product Code. 10209523 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
275 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10209523 275 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price
Product Code. 10209523 Supplier Thermo Scientific™ Supplier No. 23252

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27969” to get your promotional price

Reducing agent-compatible version of our popular BCA Reagent Kit to measure protein concentration in samples containing DTT or BME thiol-reductants.

This reducing agent-compatible (RAC) version of the Thermo Scientific™ Pierce BCA Protein Assay provides accurate determination of protein concentration in samples that contain dithiothreitol (DTT), 2-mercaptoethanol (BME), TCEP and other disulfide reducing agents.

The BCA Protein Assay Kit - Reducing Agent Compatible (BCA-RAC) provides all of the advantages of the original BCA Assay, plus compatibility with disulfide reducing agents at concentrations routinely used in protein sample buffers. This special adaptation of the popular Pierce BCA Protein Assay method enables accurate protein concentration measurement for samples containing DTT, 2-ME or TCEP. Available in tube and microplate formats, these reducing agent compatible (RAC) BCA Kits extend the already broad reagent compatibility of the BCA Protein Assay protocol to include nearly all types of components commonly present in protein research samples.

Highlights:

  • Compatible – assay samples that contain up to 5mM DTT, 35mM BME, or 10mM TCEP

  • BCA technology – only a slight modification of the standard BCA Protein Assay protocol (15-minute incubation with Compatibility Reagent); no precipitation steps required

  • Small samples – requires only 25μL (standard kit) or less than 10μL (microplate kit) of sample

  • Colorimetric – measure with a standard spectrophotometer or plate reader (562nm)

  • Excellent uniformity – exhibits less protein-to-protein variation than dye-binding methods

  • High linearity – linear working range for BSA equals 125 to 2000μg/mL

Includes:

BCA Reagents A and B, Compatibility Reagent, Reconstitution Buffer, and Albumin Standard (2mg/mL); Microplate kit also includes assay-compatible 96-well microplates

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Sufficient For: 980 microplate assays with 9 μL samples
• BCA Reagent A, 250 mL
• BCA Reagent B, 25 mL
• Compatibility Reagent, 48 x 10 mg
• Reconstitution Buffer, 15 mL
• Albumin Standard Ampules, 2mg/mL, 10 x 1 mL
• 96-Well Microplates, 20 plates

Store at room temperature
Description Pierce BCA Microplate Protein Assay Kit, Reducing Agent Compatible (RAC)
Assay BCA Assay
Detection Method Colorimetric
Product Line Pierce
Product Type Protein Quantitation Assay
Specificity Not Target-Specific
Sufficient For 980 Microplate Assays
For Use With (Application) Solution-based Detection, Absorbance
For Use With (Equipment) Spectrophotometer, Microplate Reader
Quantity 275 mL
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pierce Microplate BCA Protein Assay Kit - Reducing Agent Compatible
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P273-Avoid release to the environment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P391-Collect spillage.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: sodium carbonate, [2, 2'-Biquinoline]-4,4'-dicarboxylic acid, sodium salt (1:2), Sulfuric acid copp

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.