missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Blaubrand™ Class A Borosilicate Glass Volumetric Flasks with Glass Stopper
Constructed of Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Blaubrand™ Class A Volumetric Flask with Glass Stopper is available in both narrow neck and wide neck options in a variety of sizes. DE-M marking on neck provides quick and accurate volume check.
42.00 € - 161.00 €
Specifications
Calibration | Calibrated To Contain |
---|---|
Certifications/Compliance | DIN EN ISO 1042 |
Closure Material | Glass |
Color | Clear |
Description | DE-M Marking |
Product Code | Brand | Capacity (Metric) | Neck Style | Closure Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Neck Style | Closure Size | Price | Quantity & Availability | |||
10759852
|
BRAND™
37264 |
1000 mL | Narrow | 24/29 |
63.83 €
Each |
In Stock
Log in to see stock. |
|||
11834020
|
BRAND™
37289 |
100 mL | Narrow | 14/23 |
51.70 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
11718148
|
BRAND™
37267 |
10 mL | Wide | 10/19 |
43.65 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
11643209
|
BRAND™
37288 |
50 mL | Wide | 14/23 |
124.87 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
11824020
|
BRAND™
37258 |
25 mL | Narrow | 10/19 |
43.90 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
11868361
|
BRAND™
37256 |
5 mL | Wide | 10/19 |
42.00 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
11814020
|
BRAND™
37257 |
20 mL | Narrow | 10/19 |
43.20 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
11844120
|
BRAND™
37268 |
20 mL | Wide | 12/21 |
45.30 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
12085312
|
BRAND™
37262 |
250 mL | Narrow | 14/23 |
67.15 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
10004241
|
BRAND™
37265 |
2000 mL | Narrow | 29/32 |
85.09 €
Each |
In Stock
Log in to see stock. |
|||
10315381
|
BRAND™
37266 |
5 L | Narrow | 34/35 |
161.00 €
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||
11834120
|
BRAND™
37263 |
500 mL | Narrow | 19/26 |
69.00 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
11898361
|
BRAND™
37261 |
200 mL | Narrow | 14/23 |
55.80 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
11888361
|
BRAND™
37260 |
100 mL | Narrow | 12/21 |
49.70 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
11878361
|
BRAND™
37259 |
50 mL | Narrow | 12/21 |
47.00 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
12095312
|
BRAND™
37294 |
25 mL | Wide | 12/21 |
47.01 €
Pack of 2 |
In Stock
Log in to see stock. |
|||
Description
- Class A Volumetric Flask
- Constructed of Borosilicate Glass
- Complies with DIN EN ISO 1042 standards
- DAkkS calibration certificate available upon request
- Calibrated to contain (TC, In)
- Narrow neck and wide neck options available in a variety of sizes
- DE-M marking provides quick and accurate volume check
- Glass stopper included
Specifications
Calibrated To Contain | |
Glass | |
DE-M Marking | |
Yes |
DIN EN ISO 1042 | |
Clear | |
Borosilicate Glass | |
A |