missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ P2 Primary Cell 4D-Nucleofector™ X Kit L

Product Code. 13489319
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
12 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13489319 1 Kit 12 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13489319 Supplier Lonza™ Supplier No. V4XP2012
  • ONLINE EXCLUSIVE PRICE
    477.00 €
    397.28 € / 12 reactions
    Save 79.72 €
    16% Off
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For efficient transfection of specific primary cells, e.g., mouse macrophages and NHDF, in the 4D-NucleofectorTM X Unit (100 μL format).

  • High transfection efficiencies for primary cells.
  • Cells maintain functionality post transfection.
  • Conditions are transferable between 4D-Nucleofector™ System, 96-well Shuttle™ System and 384-well Nucleofector™ System.
  • Applications:
    • Transfection of cell numbers in the range of 2 x 105 to 2 x 107 cells.
    • Low throughput applications.
  • This kit has been determined to be optimal for transfecting following cell types:
    • Macrophage (mouse).
    • NHDF (human), neonatal.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Vector pmaxGFP™
For Use With (Equipment) 4D-Nucleofector™ X Unit (100 μL format)
For Use With (Application) Efficient transfection of specific primary cells, e.g., mouse macrophages and NHDF.
Includes P2 Primary Cell Nucleofector™ Solution, Supplement 1, pmaxGFP™ Vector (1 μg/μL in 10 mM Tris pH 8.0), Nucleocuvette™ Vessel, Single-use pipettes
No. of Reactions 12
pH 7.3 to 7.5
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.