missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals p21-Activated Kinase Inhibitor III, IPA-3

Product Code. 15497099
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15497099 10 mg 10mg
15487099 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15497099 Supplier Thermo Scientific Chemicals Supplier No. J65848.MA
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

Acts as an allosteric inhibitor of Pak1

Acts as an allosteric inhibitor of Pak1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42521-82-4
Molecular Formula C20H14O2S2, C20H14O2S2
Synonym PAK Inhibitor III; 1,1′-Disulfanediylbis(naphthalen-2-ol)

Specifications

Quantity 10 mg
Description Yellow
Content And Storage -30°C to -10°C
For Use With (Application) Acts as an allosteric inhibitor of Pak1
Formula Weight 350.50g/mol
Sensitivity Light Sensitive
Solubility Soluble in DMSO or ethanol; Insoluble in water
Product Type Signal Transduction Reagent
Form Solid
InChI Key RFAXLXKIAKIUDT-UHFFFAOYSA-N
SMILES OC1=CC=C2C=CC=CC2=C1SSC1=C(O)C=CC2=CC=CC=C12
IUPAC Name 1-[(2-hydroxynaphthalen-1-yl)disulfanyl]naphthalen-2-ol
Molecular Weight (g/mol) 350.45
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • p21-Activated Kinase Inhibitor III, IPA-3
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1,1-Dithiodi-2-naphthol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.