Learn More
PAC 3500 Hydrogen Sulphide Personal Gas Monitor
Product Code. 12314599
Change view
Click to view available options
Measurement Range:
0 to 100ppm
Unit Size:
Each
Product Code. Measurement Range Unit Size
12314599 0 to 100ppm Each
1 options
Product Code. 12314599 Supplier Draeger Safety Supplier No. 8322003
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Alarm Types Audible, visual and vibrating
Type Hydrogen Sulphide Personal Gas Monitor
Measurement Range 0 to 100ppm
Warranty 2Years