missing translation for 'onlineSavingsMsg'
Learn More

Pachymic Acid, TRC

Product Code. 30427218
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30427218 10 mg
30422722 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30427218 Supplier Toronto Research Chemicals Supplier No. TRCP13220010MG
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29070-92-6
Molecular Formula C33 H52 O5
Molecular Weight (g/mol) 528.76
Synonym Lanost-8-en-21-oic acid, 3-(acetyloxy)-16-hydroxy-24-methylene-, (3β,16α)-, Eburica-8,24(28)-dien-21-oic acid, 3β,16α-dihydroxy-, 3-acetate (6CI), Lanost-8-en-21-oic acid, 3β,16α-dihydroxy-24-methylene-, 3-acetate (8CI), (3β,16α)-3-(Acetyloxy)-16-hydroxy-24-methylenelanost-8-en-21-oic acid, 3-O-Acetyltumulosic acid, NSC 244427, Pachymic acid
IUPAC Name (2R)-2-[(3S,5R,10S,13R,14R,16R,17R)-3-acetyloxy-16-hydroxy-4,4,10,13,14-pentamethyl-2,3,5,6,7,11,12,15,16,17-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-6-methyl-5-methylideneheptanoic acid
SMILES CC(C)C(=C)CC[C@H]([C@H]1[C@H](O)C[C@@]2(C)C3=C(CC[C@]12C)[C@@]4(C)CC[C@H](OC(=O)C)C(C)(C)[C@@H]4CC3)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C33H52O5/c1-19(2)20(3)10-11-22(29(36)37)28-25(35)18-33(9)24-12-13-26-30(5,6)27(38-21(4)34)15-16-31(26,7)23(24)14-17-32(28,33)8/h19,22,25-28,35H,3,10-18H2,1-2,4-9H3,(H,36,37)/t22-,25-,26+,27+,28+,31-,32-,33+/m1/s1
Formula Weight 528.3815
Percent Purity >95%
Chemical Name or Material Pachymic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.