missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Pacific Orange™ and Pacific Blue™ Antibody Labeling Kits

Product Code. 10534453
Change view
Click to view available options
Excitation/Emission:
405/551 nm
410/455 nm
Color:
Pacific Blue
Pacific Orange
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Excitation/Emission Color Unit Size
10534453 410/455 nm Pacific Blue Each
10165212 405/551 nm Pacific Orange Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10534453 Supplier Molecular Probes™ Supplier No. P30013
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Antibody Labeling Kits specifically bind small amounts of antibody with violet light-excitable Pacific Orange or Pacific Blue dyes. Conveniently label up to 100 μg of antibody per reaction, with 5 reactions per kit.

Pacific Orange and Pacific Blue antibody labeling kits enable attachment of a violet light-excitable fluorescent label on antibodies (up to 100 μg per reaction). Each kit contains enough dye for five reactions.

A primary antibody directly labeled with a fluorophore often produces lower background fluorescence and less nonspecific binding. Primary antibodies of the same isotype or derived from the same species can be multiplexed if they are directly labeled with compatible fluorophores. Using Pacific Blue and Pacific Orange dyes simultaneously allows you to excite both dyes at the same wavelength while their emissions are differentially distinguished.

Pacific Orange and Pacific Blue antibody labeling kits allow the labeling of small amounts of antibody with a violet light-excitable dye (up to 100 μg of antibody per reaction). Small amounts of other proteins (>40 kDa) can also be labeled using these kits.

Features of these antibody labeling kits
• Pacific Blue excitation and emission maxima: 410/455 nm
• Pacific Orange excitation and emission maxima: 405/551 nm
• Labeled proteins typically ready to use in 90 min. (∼15 min. hands-on time)
• Optimized for small-scale labeling of any protein >40 kDa
• Stabilizing proteins should be removed from the sample before labeling
• Purified using convenient spin filters
• Includes detailed instructions for determining degree of labeling (DOL)

These kits contain everything you need to perform five separate labeling reactions as well as to purify the resulting conjugates. Conjugates are ideal for multiple applications, including flow cytometry, fluorescent microscopy, immunohistochemistry, primary detection, ELISA, immunocytochemistry, indirect FISH, and more. We are ISO 13485:2000 certified.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Antibody Labeling Kit
Quantity 5 Reactions kit
Content And Storage Contains 5 vials of Pacific Blue™ reactive dye, 1 vial sodium bicarbonate (84 mg), purification resin (10 mL), 5 spin columns, and and 5 collection tubes.

Store at 2-8°C, protected from light. Component A may be stored from -20°C to 8°C.
Detection Method Fluorescence
Excitation/Emission 410/455 nm
Labeling Scale 100 μg
Shipping Condition Room Temperature
Product Line Pacific Blue
Chemical Reactivity Amine
Solubility DMSO (Dimethylsulfoxide)
Label or Dye Pacific Blue
Color Pacific Blue
Labeling Target Antibodies, Proteins
Label Type Pacific Dyes
Show More Show Less
What formulation of antibody should I use for conjugation for small animal in vivo imaging?

To allow for good reaction kinetics, antibodies should be in PBS buffer at a concentration of 0.5-3.0 mg/ml. The antibody must be free of preservatives (azide etc.), amine containing buffers and carrier proteins such as BSA.

What is degree of labeling (DOL)?

Degree of labeling (DOL) describes the number of fluorophores per antibody. For in vivo labeling experiments, the DOL is restricted to a narrow range because it has significant consequences for the biodistribution and clearance of the probe. For example, for in vivo imaging, we have determined that the DOL range for the far-red Alexa Fluor dyes is 1.5 to 3 molecules per antibody for optimal optical in vivo imaging.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.