missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Paclitaxel

Product Code. 16451996
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451996 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16451996 Supplier MedChemExpress Supplier No. HYB0015
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Paclitaxel is a naturally occurring antineoplastic agent and stabilizes tubulin polymerization. Paclitaxel can cause both mitotic arrest and apoptotic cell death. Paclitaxel also induces autophagy.

Specifications

Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Molecular Weight (g/mol) 853.91 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 33069-62-4
Product Type Natural Products
Purity 98.12%
Solubility DMSO : 100 mg/mL (117.11 mM∣Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
Target Microtubule/Tubulin∣ADC Cytotoxin∣Apoptosis∣Autophagy
Quantity 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.