missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Paclitaxel, 99+%

Product Code. 10482875 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10482875 5 mg 5mg
10516261 1 mg 1mg
10093573 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10482875 Supplier Thermo Scientific Chemicals Supplier No. 328420050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Paclitaxel, also known as Taxol, CAS # 33069-62-4 is a chemotherapeutic drug that acts through stabilizing microtubule assembly and arresting cell cycle progression. | CAS: 33069-62-4 | C47H51NO14 | 853.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Paclitaxel is a naturally ocurring compound originally isolated from the tree T. brevifolia
  • Paclitaxel binds tubulin and stabilizes microtubule formation, preventing disassembly
  • Paclitaxel is used as a drug to arrest cells at the mitotic checkpoint and block cell cycle progression, thus inducing death of cancer cells

Applications

  • Paclitaxel can be used in preclinical laboratory research on diseases such as cancer, heart disease, inflammatory disorders, and hepatic fibrosis
  • Paclitaxel can be used as a tool in cell biology to study mitotic checkpoint arrest conditions and downstream signaling cascades
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33069-62-4
Molecular Formula C47H51NO14
Molecular Weight (g/mol) 853.92
MDL Number MFCD00869953
InChI Key RCINICONZNJXQF-VAZQATRQSA-N
Synonym paclitaxel, taxol, taxol a, abraxane, paxene, paxceed, plaxicel, yewtaxan, onxol, ebetaxel
PubChem CID 133640187
IUPAC Name (1S,2S,4S,7R,9S,10S,12R,15S)-4,12-bis(acetyloxy)-1,9-dihydroxy-15-{[(2R,3S)-2-hydroxy-3-phenyl-3-(phenylformamido)propanoyl]oxy}-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate
SMILES CC(=O)O[C@@H]1C2=C(C)[C@H](C[C@@](O)([C@@H](OC(=O)C3=CC=CC=C3)C3[C@@]4(CO[C@@H]4C[C@H](O)[C@@]3(C)C1=O)OC(C)=O)C2(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 213.0°C to 216.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Quantity 5 mg
Merck Index 15, 7081
Solubility Information Solubility in water: sparingly soluble. Other solubilities: soluble in methanol,dmso,acetonitrile
Formula Weight 853.9
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Paclitaxel, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Paclitaxel
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H361-Suspected of damaging fertility or the unborn child.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.