missing translation for 'onlineSavingsMsg'
Learn More

Kern Sohn Padded Wooden Box for Weights

Product Code. 17352173
Encompass_Preferred
Click to view available options
Holds:
10 kg
20 kg
50 kg
Classification:
F1
F2, M1
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17352173

Brand: Kern Sohn 337141100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Wooden case for standard weight sets.

  • Wooden
  • For check weight design

Specifications

Product Type Case
Classification F1
Case Material Wood
For Use With (Equipment) Check Weights
Holds 10 kg
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.