Learn More
DiscoverX PAF
Product Code. 17382499
Encompass
Click to view available options
:
1 mg
Product Code. 17382499
Brand: DiscoverX 921179
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
PAF is an agonist for PTAFR. Provided as 1mg of dry powder.
Platelet-activating factor (β-Acetyl-γ-O-alkyl-L-α-phosphatidylcholine, or PAF) is a phospholipid that mediates many leukocyte functions. Stimulates platelet aggregation and has roles in inflammation, anaphylaxis and wound repair. Produced by lymphocytes, platelets, and endothelial cells, and is also found to be secreted by tumor cells. Acts as an agonist for PTAFR. Provided as 1mg of dry powder.
Specifications
Quantity 1 mg