missing translation for 'onlineSavingsMsg'
Learn More

Pafuramidine, TRC

Product Code. 30427764
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427764 5 mg -
30428269 10 mg -
30413487 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427764 Supplier Toronto Research Chemicals Supplier No. TRCP1338755MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 186953-56-0
Molecular Formula C20H20N4O3
Molecular Weight (g/mol) 364.4
Synonym 2,5-Bis(4-amidinophenyl)furan-bis-O-methylamidoxime, 4,4'-(2,5-Furandiyl)bis[N-methoxybenzenecarboximidamide], 4,4'-(2,5-Furandiyl)bis[N-methoxybenzenecarboximidamide
IUPAC Name N-methoxy-4-[5-[4-(N-methoxycarbamimidoyl)phenyl]furan-2-yl]benzenecarboximidamide
SMILES CONC(=N)c1ccc(cc1)c2oc(cc2)c3ccc(cc3)C(=N)NOC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C20H20N4O3/c1-25-23-19(21)15-7-3-13(4-8-15)17-11-12-18(27-17)14-5-9-16(10-6-14)20(22)24-26-2/h3-12H,1-2H3,(H2,21,23)(H2,22,24)
Formula Weight 364.1535
Chemical Name or Material Pafuramidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.