missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PageRuler™ Unstained Broad Range Protein Ladder

Product Code. 17116108 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 250 μL
8 x 250 μL
Unit Size:
Pack of 2
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17116108 8 x 250 μL Pack of 4
11892124 2 x 250 μL Pack of 2
15563197 8 x 250 μL Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17116108 Supplier Thermo Scientific™ Supplier No. 26630X4
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A mixture of 11 proteins (5 to 250kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.

Thermo Scientific™ PageRuler Unstained Broad Range Protein Ladder is a mixture of 11 proteins (5 to 250kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and Western blotting.

The protein MW markers in this ladder resolve into clearly identifiable sharp bands ranging in size from 5kDa to 250kDa when analyzed by SDS-PAGE and stained with coomassie dye or silver. The 100, 50 and 20kDa protein bands are of greater intensity and serve as reference bands. Proteins can be detected on Western blots by staining with Ponceau S, coomassie blue or other protein stains. In addition, the ladder proteins (except for the 5kDa peptide) contain an integral Strep-tag™ II Sequence and may be detected on Western blots using Strep-Tactin™ Conjugates. The PageRuler Unstained Broad Range Protein Ladder is supplied ready-to-use: no heating, dilution or addition of a reducing agent is required before use.

Highlights:

  • Size range – eleven proteins spanning 5 to 250kDa
  • Ready-to-use – supplied in a loading buffer for direct loading on gels; no need to boil
  • Sharp bands – excellent accuracy for detection in with coomassie or silver stains
  • Reference bands – the 100, 50 and 20kDa bands are more intense for easy orientation
  • Tagged – each protein in the ladder contains an integral Strep-tag™ II Sequence
  • Quality tested – each lot evaluated by SDS-PAGE and Western blotting

Includes:

Proteins in 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 100 mM DTT, 1 mM NaN3, 0.01% bromophenol blue and 33% glycerol.

Recommended for:

Accurate protein sizing on SDS-polyacrylamide gels and Western blots

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Contents: eight vials of 250 μL each

Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3, 0.01% bromophenol blue and 33% glycerol

Storage: Upon receipt store at -20°C

Number of Markers 11
Ready to Load Yes
Size Range 5 to 250 kDa
Molecular Weight (g/mol) 250, 150, 100, 70, 50, 40, 30, 20, 15, 10, 5 kDa
Quantity 8 x 250 μL
Product Line PageRuler
Product Type Protein Ladder
Stain Type Unstained
System Type SDS-PAGE

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.