missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Paint

Product Code. 11741106 Shop All Brady Products
Change view
Click to view available options
Color:
Black
White
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11741106 Black Each
11731106 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11741106 Supplier Brady™ Supplier No. 804319
  • ONLINE EXCLUSIVE PRICE
    244.00 €
    232.45 € / Pack of 1
    Save 11.55 €
    4% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Paint for floor and road markings.

  • For Floor Marking.
  • For Traffic Control.

Specifications

Quantity 1 Ea.
Alternate Part Number PEINTURE ROUTIERE (5KG) NOIR
Product Type Paint
For Use With (Application) Floor Marking, Traffic Control
Color Black
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.