missing translation for 'onlineSavingsMsg'
Learn More

Atago™ PAL-88S Model Digital Hand-Held Pocket Refractometer

Product Code. 10091102
Change view
Click to view available options
Temperature Range (Metric):
10°C to 40°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) unitSize
10091102 10°C to 40°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10091102 Supplier Atago™ Supplier No. 4488
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Type Handheld Digital Pocket Refractometer
Scale Range 0 to 90% (V/V) Propylene Glycol, 0°C to -50°C Propylene Glycol Freezing Point
Model PAL-88S
Accuracy ±0.4% (V/V) Propylene Glycol/±1°C Freezing Point of Propylene Glycol/±1°C Temperature
Resolution 0.2% (V/V) Propylene Glycol Concentration/1°C Freezing Point of Propylene Glycol/0.1°C Temperature
Measurement Time 3 sec.
IP Rating IP65
Weight (Metric) 100 g
Dimensions (L x W x H) 31 x 55 x 109 mm
Height (Metric) 109 mm
Length (Metric) 55 mm
Width (Metric) 31 mm
Ambient Temperature (Metric) 10°C to 40°C
Temperature Range (Metric) 10°C to 40°C
Power Supply Details 2xAAA Batteries
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.