missing translation for 'onlineSavingsMsg'
Learn More

PAL International™ PAL TX Surface Disinfectant PLUS Wipe Bucket

Product Code. 15864477
Change view
Click to view available options
Dimensions (L x W):
195 x 235 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15864477 195 x 235 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15864477 Supplier PAL International™ Supplier No. W64230T
  • ONLINE EXCLUSIVE PRICE
    35.00 €
    34.63 € / Pack of 1
    Save 0.37 €
    1% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for heavy duty cleaning and disinfecting of large food contact surfaces

  • These strong, cloth-like wipes are designed to deliver the same, highly effective food safe disinfectant on a high performance material
  • Pal TX Surface Disinfectant wipes are Food safe and proven to kill most microorganisms such as bacteria, yeast & viruses
  • Pal TX Surface Disinfectant Wipes clean and disinfect without using Quats, PHMB or alcohol

Specifications

Dimensions (L x W) 195 x 235 mm
Includes 8 L bucket with 500 PLUS wipes
compliance-icons
Product Identifier
  • Pal TX Surface Disinfectant Wipes
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: N-(3-aminopropyl)-N-dodecylpropane-1,3-diamine, isotridecanol ethoxylated, alcohols, C12-14, ethoxyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.