missing translation for 'onlineSavingsMsg'
Learn More

PAL International™ PAL TX Surface Disinfectant Wipes

Product Code. 15854477
Change view
Click to view available options
Dimensions (L x W):
130 x 130 mm
Unit Size:
Each
Product Code. Dimensions (L x W) unitSize
15854477 130 x 130 mm Each
1 options
This item is not returnable. View return policy

Product Code. 15854477

Brand: PAL International™ W20230T

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Small cannister with 200 Food & Beverage surface wipes

  • 0.5 L Canister is convenient for storage
  • This is a perfect size square wipe for the disinfection of small surfaces, probes and utensils in food environments
  • Pal TX Surface Disinfectant wipes are Food safe and proven to kill most microorganisms such as bacteria, yeast & viruses
  • Pal TX Surface Disinfectant Wipes clean and disinfect without using Quats, PHMB or alcohol

Specifications

Dimensions (L x W) 130 x 130 mm
Includes 0.5 L cannister with 200 wipes
Product Identifier
  • Pal TX Surface Disinfectant Wipes
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : N-(3-aminopropyl)-N-dodecylpropane-1,3-diamine, isotridecanol ethoxylated, alcohols, C12-14, ethoxyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.