missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Pala 405/488 Single Cell Sorter and Dispenser System Bundle, EMEA

Product Code. 31781569
Change view
Click to view available options
Format:
Benchtop
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Unit Size
31781569 Benchtop 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For research use only. Operate by trained personnel and follow laboratory biosafety procedures when handling biological samples. Use only approved sheath fluids and shutdown procedures to prevent system damage.
Product Code. 31781569 Supplier R&D Systems™ Supplier No. NI007FSEMEA
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For research use only. Operate by trained personnel and follow laboratory biosafety procedures when handling biological samples. Use only approved sheath fluids and shutdown procedures to prevent system damage.

The Pala 405/488 Single Cell Sorter and Dispenser System Bundle enables gentle, microfluidics-based isolation and dispensing of single cells into plates or tubes. Integrated flow cytometry detection supports precise cell selection, high viability, and reliable clonal outgrowth for cell line development and single-cell workflows.

  • Microfluidics-based single cell sorting and dispensing platform designed for gentle cell handling and high viability
  • Integrated flow cytometry detection enables precise selection of fluorescent and non-fluorescent cell populations
  • 405 nm and 488 nm laser configuration supports expanded fluorescence detection and multicolor applications
  • Dispenses single cells directly into 96- or 384-well plates for rapid clonal isolation and monoclonal cell line development
  • Bulk sorting capability enables enrichment and collection of target cell populations into tubes
  • Low-pressure sorting (<2 psi) preserves cell integrity and supports sensitive cell types including primary and stem cells
  • Sterile, disposable microfluidic cartridges eliminate sample cross-contamination and reduce maintenance
  • High single-cell dispensing accuracy (>95%) supports reliable downstream analysis and cloning workflows
  • Compact benchtop footprint suitable for standard laboratories or biosafety cabinets
  • Fast processing enables dispensing of a 96-well plate in approximately 2 min.
  • Supports applications including cell line development, CRISPR editing, single-cell genomics, antibody discovery, and rare cell isolation
  • User-friendly software interface simplifies setup, gating, and dispensing operations
  • Bundle includes required consumables for immediate system operation
  • Designed for research use workflows requiring reproducibility, sterility, and ease of use
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Instrument system supplied with laptop workstation, sheath and waste bottles, cleaning cartridge, adapters, and accessories. Store and operate in a clean, dry laboratory environment.
Dimensions (D x W x H) 36 x 65 x 23 cm
Voltage 100/115/220/230 V
Frequency 50/60 Hz
Product Type Single Cell Sorter and Dispenser
Laser Features Dual-laser Excitation: 405 nm and 488 nm with multi-parameter fluorescence detection
Product Line Pala
Includes Pala instrument, laptop workstation with software, sheath and waste bottles, cleaning cartridge, adapters, single-cell and bulk cartridge kits
Format Benchtop
Quantity 1

For research use only. Designed for use with disposable Pala cell cartridges and compatible microtiter plates. Performance may vary depending on sample preparation and cell type.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.