missing translation for 'onlineSavingsMsg'
Learn More

Palatinose Monohydrate, TRC

Product Code. 30417444
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30417444 1 g 1g
30410811 50 g -
30412321 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30417444 Supplier Toronto Research Chemicals Supplier No. TRCP1398901G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58024-13-8
Molecular Formula C12H22O11 . H2O
Molecular Weight (g/mol) 360.32
Synonym 6-O-α-D-Glucopyranosyl-D-fructose Monohydrate, Galen IQ Hydrate, Palatinose Monohydrate, Isomaltulose Monohydrate, Palatinose IC Monohydrate, Palatinose ICP Monohydrate
IUPAC Name (3S,4R,5R)-1,3,4,5-tetrahydroxy-6-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexan-2-one hydrate
SMILES OC[C@H]1O[C@H](OC[C@H]([C@H]([C@@H](C(CO)=O)O)O)O)[C@H](O)[C@@H](O)[C@@H]1O.O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H22O11.H2O/c13-1-4(15)7(17)8(18)5(16)3-22-12-11(21)10(20)9(19)6(2-14)23-12;/h5-14,16-21H,1-3H2;1H2/t5-,6-,7-,8-,9-,10+,11-,12+;/m1./s1
Formula Weight 360.13
Chemical Name or Material Palatinose Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.