missing translation for 'onlineSavingsMsg'
Learn More

Palbociclib monohydrochloride, MedChemExpress

Product Code. 16487866
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487866 100 mg 100mg
16430568 5 mg 5mg
16428108 10 mg 10mg
16404027 50 mg 50mg
16401326 200 mg 200mg
16421926 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16487866 Supplier Medchem Express Supplier No. HY50767A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Palbociclib (PD 0332991) monohydrochloride is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. Palbociclib monohydrochloride has potent anti-proliferative activity and induces cell cycle arrest in cancer cells, which can be used in the research of HR-positive and HER2-negative breast cancer and hepatocellular carcinoma.

Palbociclib (PD 0332991) monohydrochloride is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. Palbociclib monohydrochloride has potent anti-proliferative activity and induces cell cycle arrest in cancer cells, which can be used in the research of HR-positive and HER2-negative breast cancer and hepatocellular carcinoma.

Specifications

Chemical Name or Material Palbociclib monohydrochloride
CAS 827022-32-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H30ClN7O2
Quantity 100 mg
Synonym PD 0332991 monohydrochloride
Solubility Information H2O : 7.14 mg/mL (14.75 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 4.63 mg/mL (9.57 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C1C(C(C)=O)=C(C)C2=CN=C(NC3=NC=C(N4CCNCC4)C=C3)N=C2N1C5CCCC5.Cl
Molecular Weight (g/mol) 483.99
Formula Weight 483.99
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.