missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Paliperidone

Product Code. 18772524
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18772524 10 mg 10mg
18712684 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18772524 Supplier R&D Systems™ Supplier No. 4493/10
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Atypical antipsychotic; 5-HT2A and D2 antagonist

Atypical antipsychotic; metabolite of risperidone (Cat. No. 2865). Exhibits high affinity for 5-HT2A receptors (Ki = 0.4 nM for cloned human 5-HT2A receptors). Also displays nanomolar affinity for human D2 receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144598-75-4
Molecular Formula C23H27FN4O3

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 426.48
Purity >98%
Molecular Weight (g/mol) 426.492
Chemical Name or Material Paliperidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.