missing translation for 'onlineSavingsMsg'
Learn More

Paliperidone Palmitate, TRC

Product Code. 30011018
Change view
Click to view available options
Quantity:
10 mg
250 mg
1 g
Unit Size:
10mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011018 10 mg 10mg
30006787 1 g 1g
30003610 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011018 Supplier Toronto Research Chemicals Supplier No. TRCP14102010MG
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 199739-10-1
Molecular Formula C39 H57 F N4 O4
Molecular Weight (g/mol) 664.89
Synonym Hexadecanoic acid, 3-[2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidin-9-yl ester, 9-Hydroxyrisperidone palmitate, Invega Sustenna, Paliperidone palmitate, RO 92670, Hexadecanoic acid 3-[2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidin-9-yl ester, [2-Methyl-4-oxo-3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-9-yl] hexadecanoate
IUPAC Name [3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-4-oxo-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-9-yl] hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC1CCCN2C(=O)C(=C(C)N=C12)CCN3CCC(CC3)c4noc5cc(F)ccc45

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C39H57FN4O4/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-18-36(45)47-34-17-16-24-44-38(34)41-29(2)32(39(44)46)23-27-43-25-21-30(22-26-43)37-33-20-19-31(40)28-35(33)48-42-37/h19-20,28,30,34H,3-18,21-27H2,1-2H3
Formula Weight 664.4364
Chemical Name or Material Paliperidone Palmitate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.