missing translation for 'onlineSavingsMsg'
Learn More

Palladium on 2-4 mm alumina pellets, 0.5% Pd

Product Code. 10431241 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10431241 50 g Glass bottle 50g
10226730 250 g Glass bottle 250g
11926951 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10431241 Supplier Thermo Scientific Acros Supplier No. 195080500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7440-05-3 | Pd | 106.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5-3-7440
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Color Gray
Density 700g/L (tapped)
Quantity 50 g
Packaging Glass bottle
Form Pellet
Fieser 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Formula Weight 106.42
Percent Purity 0.5% (Pd)
Chemical Name or Material Palladium on 1/8-inch alumina pellets

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.