missing translation for 'onlineSavingsMsg'
Learn More

Palladium (5% on Barium Sulfate), TRC

Product Code. 30412322
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30412322 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412322 Supplier Toronto Research Chemicals Supplier No. TRCP1410505G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Palladium
CAS 7440-05-3
Molecular Formula Pd
Quantity 5 g
Synonym Palladium on carbon, Palladium Black, Paladio, Palladium/carbon, Palladium, IG 0218A, MPP 030, MPP 050, MPP 080, P 50, P 50 (metal), Palladex 600, Palladium (nanoparticles), Palladium (powder), Palladium element, SFP 1001P, TCP 211
InChI Formula InChI=1S/Pd
SMILES [Pd]
IUPAC Name palladium
Molecular Weight (g/mol) 106.42
Formula Weight 105.9035
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.