missing translation for 'onlineSavingsMsg'
Learn More

Palladium (II) Chloride, TRC

Product Code. 30416021
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416021 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30416021 Supplier Toronto Research Chemicals Supplier No. TRCP1411502.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Palladium (II) Chloride
CAS 7647-10-1
Molecular Formula PdCl2
Quantity 2.5 g
Synonym Dichloropalladium, Enplate Activator 440, NSC 146183, Niklad 262, Palladium Dichloride, Palladous Chloride
InChI Formula InChI=1S/2ClH.Pd/h2*1H;/q;;+2/p-2
SMILES Cl[Pd]Cl
IUPAC Name dichloropalladium
Molecular Weight (g/mol) 177.33
Formula Weight 175.8412
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.