missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) iodide, Premion™, 99.998% (metals basis), Pd 29% min

Product Code. 11334738
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11334738

Brand: Thermo Scientific Alfa Aesar 043084.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-38-7 | I2Pd | 360.23 g/mol

Used as catalyst. Also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-38-7
Molecular Formula I2Pd
Molecular Weight (g/mol) 360.23
MDL Number MFCD00011171
InChI Key HNNUTDROYPGBMR-UHFFFAOYSA-L
IUPAC Name palladium(2+) diiodide
SMILES [Pd++].[I-].[I-]

Specifications

Melting Point 350°C (decomposition)
Color Black
Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in H2O and organic solvents. Insoluble in diluted nitric acid and hydrochloric acid. Slightly soluble in hydriodic acid and hot concentrated nitric acid. Soluble in ammonia and methyl acetate.
Formula Weight 360.23
Percent Purity 99.998%
Grade Premion™
Concentration or Composition (by Analyte or Components) Pd 29% min
Density 6.003 g/mL
Assay (metals basis)
Chemical Name or Material Palladium (II) iodide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.