missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) nitrate hydrate

Product Code. 10279243 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10279243 1 g Glass bottle 1g
10239293 5 g Glass bottle 5g
11994361 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10279243 Supplier Thermo Scientific Acros Supplier No. 389910010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207596-32-5 | N2O6Pd | 230.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207596-32-5 , 10102-05-3
Molecular Formula N2O6Pd
Molecular Weight (g/mol) 230.43
MDL Number MFCD00149818
InChI Key GPNDARIEYHPYAY-UHFFFAOYSA-N
Synonym palladium ii nitrate hydrate, palladium nitrate hydrate, palladium 2+ hydrate dinitrate, acmc-1bnae, palladous nitrate hydrate, palladium dinitrate hydrate, 2no3.pd.h2o, ksc492s0r, palladium ii nitrate,hydrate, palladium,diaquabis nitrato-o-9ci
PubChem CID 16211503
SMILES [Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Black-Brown
Physical Form Crystals
Quantity 1 g
Linear Formula Pd(NO3)2·xH2O
IUPAC Name palladium(2+) dinitrate
Formula Weight 230.43
Packaging Glass bottle
Chemical Name or Material Palladium(II) nitrate hydrate
compliance-icons compliance-icons
Product Identifier
  • Palladium(II) nitrate hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.