missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) oxide, 99.995%, (trace metal basis)

Product Code. 10804161 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10804161 1 g Glass bottle 1g
10411820 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10804161 Supplier Thermo Scientific Acros Supplier No. 193680010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

CAS: 1314-08-5 | OPd | 122.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1314-08-5
Molecular Formula OPd
Molecular Weight (g/mol) 122.42
MDL Number MFCD00011172
InChI Key JQPTYAILLJKUCY-UHFFFAOYSA-N
Synonym palladium ii oxide, palladium oxide pdo, acmc-1bqvh, palladium oxide, pdo, palladium ii oxide, anhydrous, palladium ii oxide-pd, 11113-77-2 cpd with unspecified mf, palladium oxide ? inverted exclamation mark-?, palladium ii oxide trace metals basis
PubChem CID 5083724
IUPAC Name palladium(2+) oxidandiide
SMILES [O--].[Pd++]

Especificaciones

Melting Point 870.0°C
Physical Form Viscous Liquid
Quantity 1 g
Assay Percent Range 85.2 to 88.7% (Pd)
Linear Formula PdO
Merck Index 15,7094
Solubility Information Solubility in water: practically insoluble. Other solubilities: practically insoluble in acids, slightly soluble in aqua regia, soluble in 48% hbr
Formula Weight 122.4
Percent Purity 99.995%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Palladium(II) oxide
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.