missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) trifluoroacetate, 97%

Product Code. 10247760 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10247760 1 g Glass bottle 1g
10782954 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10247760

Brand: Thermo Scientific Acros 317020010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 42196-31-6 | C4F6O4Pd | 332.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42196-31-6
Molecular Formula C4F6O4Pd
Molecular Weight (g/mol) 332.45
MDL Number MFCD00013204
InChI Key PBDBXAQKXCXZCJ-UHFFFAOYSA-L
Synonym palladium ii trifluoroacetate, palladium ii 2,2,2-trifluoroacetate, palladium 2+ ion ditrifluoroacetate, palladium trifluoroacetate, palladium 2+ ditrifluoroacetate, trifluoroacetic acid palladium ii salt, bis 2,2,2-trifluoroacetoxy palladium, acmc-209jo7, ksc235m3d
PubChem CID 2733347
IUPAC Name palladium(2+);2,2,2-trifluoroacetate
SMILES [Pd++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F

Specifications

Melting Point 210°C
Color Blue to Blue-Green
Infrared Spectrum Authentic
Assay Percent Range 30.9 to 33.2% (Pd)
Packaging Glass bottle
Linear Formula (CF3CO2)2Pd
Quantity 1 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 332.44
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Palladium(II) trifluoroacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.