missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) trifluoroacetate, 97%

Product Code. 10782954 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10782954 5 g Glass bottle 5g
10247760 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10782954 Supplier Thermo Scientific Acros Supplier No. 317020050
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 42196-31-6 | C4F6O4Pd | 332.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42196-31-6
Molecular Formula C4F6O4Pd
Molecular Weight (g/mol) 332.45
MDL Number MFCD00013204
InChI Key PBDBXAQKXCXZCJ-UHFFFAOYSA-L
Synonym palladium ii trifluoroacetate, palladium ii 2,2,2-trifluoroacetate, palladium 2+ ion ditrifluoroacetate, palladium trifluoroacetate, palladium 2+ ditrifluoroacetate, trifluoroacetic acid palladium ii salt, bis 2,2,2-trifluoroacetoxy palladium, acmc-209jo7, ksc235m3d
PubChem CID 2733347
IUPAC Name palladium(2+);2,2,2-trifluoroacetate
SMILES [Pd++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F

Specifications

Melting Point 210°C
Color Blue to Blue-Green
Infrared Spectrum Authentic
Assay Percent Range 30.9 to 33.2% (Pd)
Packaging Glass bottle
Linear Formula (CF3CO2)2Pd
Quantity 5 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 332.44
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Palladium(II) trifluoroacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.