missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) Trifluoroacetate, TRC

Product Code. 30413473
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30413473 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30413473

Brand: Toronto Research Chemicals TRCP1411551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Palladium(II) Trifluoroacetate
CAS 42196-31-6
Molecular Formula 2(C2 H F3 O2 ) . Pd
Quantity 1 g
Synonym Palladium Bis(2,2,2-trifluoroacetate), Palladium Bis(trifluoroacetate), Palladium Trifluoroacetate, Trifluoroacetic Acid palladium(2+) Salt Trifluoroacetic Acid Palladium(II) Salt
InChI Formula InChI=1S/2C2HF3O2.Pd/c2*3-2(4,5)1(6)7;/h2*(H,6,7);
SMILES O=C(C(F)(F)F)O.O=C(C(F)(F)F)O.[Pd]
IUPAC Name Palladium(II) trifluoroacetate
Molecular Weight (g/mol) 334.46
Formula Weight 333.88
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.