missing translation for 'onlineSavingsMsg'
Learn More

Palladium Nickel gauze, 50 mesh woven from 0.1578mm (0.0062in) dia wire

Product Code. 11449920
Change view
Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449920 150 x 150 mm Each
11439920 75 x 75 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11449920 Supplier Thermo Scientific Alfa Aesar Supplier No. 044698.GT
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Filtration

Palladium Nickel gauze is used in filtrations. It is also used as a capture gauze for platinum recovery in ammonia oxidation apparatus with a pressure of 4 atm (405 kPa) and a temperature of 8500C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Palladium Nickel gauze
Name Note 50 mesh woven from 0.1578mm (0.0062 in.) dia. wire
Avg. Mol. Wt. or Mol. Wt. Range Pd:Ni; 95:5 wt%
Quantity 150 x 150 mm
Mesh Size 50 Mesh
Form Wire Cloth
MDL Number MFCD05863544
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.