missing translation for 'onlineSavingsMsg'
Learn More

Palladium Thinfoil, 0.0125mm (0.0005in) thick, 99.95% (metals basis)

Product Code. 11460331
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
25mm
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460331 50 x 50 mm Each
11308668 25 x 25 mm 25mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460331 Supplier Thermo Scientific Alfa Aesar Supplier No. 040744.FI
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-05-3 | Pd | 106.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5-3-7440
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Quantity 50 x 50 mm
Odor Odorless
Weight ≈0.094g/25x25mm
Form Foil
Percent Purity 99.95%
Assay (metals basis)
Chemical Name or Material Palladium Thinfoil, 0.0125mm (0.0005 in.) thick, Not certified pinhole free

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.