missing translation for 'onlineSavingsMsg'
Learn More

Palmatine chloride, 97%, Thermo Scientific™

Product Code. 17565535 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
17565535 1 g 1g
17575535 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 17565535

Brand: Thermo Scientific Acros 467150010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 10605-02-4
Molecular Formula C21 H22 Cl N O4

Specifications

Quantity 1 g
Formula Weight 387.86
MDL Number MFCD00016656
Physical Form Crystalline Powder
Color Light-yellow to Yellow to Orange
Purity ≥97.5%
InChI Key RLQYRXCUPVKSAW-UHFFFAOYSA-M
SMILES [Cl-].COC1=CC=C2C=C3C4=CC(OC)=C(OC)C=C4CC[N+]3=CC2=C1OC
IUPAC Name 3,4,10,11-tetramethoxy-7,8-dihydro-6λ⁵-azatetraphen-6-ylium chloride
Molecular Weight (g/mol) 387.86
Chemical Name or Material Palmatine chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.